Diphenylcyclopropenone

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2,3-Diphenyl-2-cyclopropen-1-one, 2,3-Diphenylcycloprop-2-en-1-one, 2,3-Diphenyl-2-cyclopropen-1-on, 1,2-Diphenylcyclopropen-3-one

OR22091-5G OR22091-1G
76570-522EA 78.44 USD
76570-522 76570-520
Diphenylcyclopropenone
Diphenylcyclopropenone
Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 407
Melting Pt: 118…122 °C
Storage Temperature: Ambient
MDL Number: MFCD00001311
CAS Number: 886-38-4
EINECS: 212-948-4
Merck Index: 12,03366

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR