(3-Methoxyphenyl)acetic acid ethyl ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Ethyl (3-methoxyphenyl)acetate, Ethyl 2-(3-methoxyphenyl)acetate

OR21783-5G OR21783-25G
76570-344EA 23.44 USD
76570-344 76570-346
(3-Methoxyphenyl)acetic acid ethyl ester
(3-Methoxyphenyl)acetic acid ethyl ester
Formula: C₁₁H₁₄O₃
Boiling Pt: 109…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00026897
CAS Number: 35553-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR