4-Chloro-3-iodophenol

Supplier: APOLLO SCIENTIFIC
Warning

OR2164-1G OR2164-10G
76570-310EA 27 USD
76570-310 76570-312
4-Chloro-3-iodophenol
4-Chloro-3-iodophenol
Formula: C₆H₄ClIO
MW: 254.45 g/mol
Melting Pt: 75…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00070769
CAS Number: 202982-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR