2,3-Dimethylmaleic anhydride 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3,4-Dimethylfuran-2,5-dione

OR21481-5G OR21481-25G OR21481-100G
76570-266EA 20.77 USD
76570-266 76570-268 76570-270
2,3-Dimethylmaleic anhydride 98%
2,3-Dimethylmaleic anhydride
Formula: C₆H₆O₃
Boiling Pt: 222…223 °C
Melting Pt: 93…96 °C
Storage Temperature: Ambient
MDL Number: MFCD00005523
CAS Number: 766-39-2
EINECS: 212-165-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR