3,4-Dihydroxy-5-methoxybenzaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR2136-5G OR2136-1G
76570-200EA 96.78 USD
76570-200 76570-198
3,4-Dihydroxy-5-methoxybenzaldehyde
3,4-Dihydroxy-5-methoxybenzaldehyde
Formula: C₈H₈O₄
Melting Pt: 131…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00016610
CAS Number: 3934-87-0
EINECS: 223-513-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR