6-Chloroanthranilic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-6-chlorobenzoic acid

OR21294-25G OR21294-100G OR21294-5G
76570-178EA 13.33 USD
76570-178 76570-180 76570-182
6-Chloroanthranilic acid
6-Chloroanthranilic acid
Formula: C₇H₆ClNO₂
MW: 171.58 g/mol
Melting Pt: 142…145 °C
Storage Temperature: Ambient
MDL Number: MFCD00051530
CAS Number: 2148-56-3
EINECS: 218-416-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR