4-Hydroxy-3,5-dimethylbenzaldehyde
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3,5-Dimethyl-4-hydroxybenzaldehyde
OR21230-500G
OR21230-100G
76570-160EA
13
USD
76570-160
76570-162
4-Hydroxy-3,5-dimethylbenzaldehyde
4-Hydroxy-3,5-dimethylbenzaldehyde
Formula:
C₉H₁₀O₂ MW: 150.18 g/mol Melting Pt: 112…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006946 CAS Number: 2233-18-3 EINECS: 218-774-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...