2-Methoxycinnamic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR21169-25G OR21169-5G
76570-146EA 23.42 USD
76570-146 76570-144
2-Methoxycinnamic acid 98%
2-Methoxycinnamic acid
Formula: C₁₀H₁₀O₃
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00064238
CAS Number: 6099-03-2
EINECS: 228-047-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR