3-(3-Bromophenyl)-1H-pyrazole
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3-(3-Bromophenyl)-1H-pyrazole
OR20-1G
OR20-5G
76569-912EA
249
USD
76569-912
76569-910
3-(3-Bromophenyl)-1H-pyrazole
3-(3-Bromophenyl)-1H-pyrazole
Formula:
C₉H₇BrN₂ MW: 223.07 g/mol Melting Pt: 73…74 °C Storage Temperature: Ambient |
MDL Number:
MFCD01940433 CAS Number: 149739-65-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...