Methyl-1-cyclopentene-1-carboxylate
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
Methyl 1-cyclopentene-1-carboxylate
OR1990-10G
OR1990-25G
OR1990-5G
76569-904EA
234
USD
76569-904
76569-902
76569-900
Methyl-1-cyclopentene-1-carboxylate
Methyl 1-cyclopentene-1-carboxylate
Formula:
C₇H₁₀O₂ MW: 126.1531 g/mol Boiling Pt: 76…78 °C Density: 1.031 g/cm³ (25 °C) Flash Pt: 62 °C Storage Temperature: Ambient |
MDL Number:
MFCD00239506 CAS Number: 25662-28-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...