2-Hydroxyisonicotinic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Hydroxy-4-pyridinecarboxylic acid, 2-Hydroxy-isonicotinic acid

OR1910-25G OR1910-100G OR1910-5G
76569-720EA 44.64 USD
76569-720 76569-722 76569-718
2-Hydroxyisonicotinic acid
2-Hydroxyisonicotinic acid
Formula: C₆H₅NO₃
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318566
CAS Number: 22282-72-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR