6-Bromopicolinic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid

OR1906-25G OR1906-5G
76569-700EA 34.64 USD
76569-700 76569-698
6-Bromopicolinic acid
6-Bromopicolinic acid
Formula: C₆H₄BrNO₂
MW: 202.01
Melting Pt: 188…194 °C
Storage Temperature: Ambient
MDL Number: MFCD00093197
CAS Number: 21190-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR