2-Bromoisonicotinic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-4-pyridinecarboxylic acid, 2-Bromopyridine-4-carboxylic acid

OR1905-25G OR1905-5G
76569-696EA 41.76 USD
76569-696 76569-694
2-Bromoisonicotinic acid 97%
2-Bromoisonicotinic acid
Formula: C₆H₄BrNO₂
MW: 202.01
Melting Pt: 217…229 °C
Storage Temperature: Ambient
MDL Number: MFCD01646069
CAS Number: 66572-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR