3-Quinolinecarbonitrile
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3-Cyanoquinoline
OR18815-5G
OR18815-1G
OR18815-10G
76569-640EA
196
USD
76569-640
76569-638
76569-636
3-Quinolinecarbonitrile
3-Quinolinecarbonitrile
Formula:
C₁₀H₆N₂ MW: 154.17 g/mol Melting Pt: 108…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006766 CAS Number: 34846-64-5 EINECS: 252-248-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...