2,3-Dimethoxybenzylamine, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-(2,3-Dimethoxyphenyl)methanamine, (2,3-Dimethoxyphenyl)methanamine

OR18556-25G OR18556-5G
76569-502EA 128.36 USD
76569-502 76569-500
2,3-Dimethoxybenzylamine, Technical Grade
2,3-Dimethoxybenzylamine
Formula: C₉H₁₃NO₂
MW: 167.21
Boiling Pt: 137 °C/11mmHg
Density: 1.13 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00052392
CAS Number: 4393-09-3
EINECS: 224-515-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR