L(-)-Carnitine 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR18381-25G OR18381-100G
76569-444EA 104.03 USD
76569-444 76569-440 76569-442
L(-)-Carnitine 98%
L(-)-Carnitine
Formula: C₇H₁₅NO₃
Melting Pt: 210…212 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00038747
CAS Number: 541-15-1
EINECS: 208-768-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR