Methyl 3-amino-6-iodobenzothiophene-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR183429-250MG OR183429-1G
76569-302EA 124.28 USD
76569-302 76569-304
Methyl 3-amino-6-iodobenzothiophene-2-carboxylate
Methyl 3-amino-6-iodobenzothiophene-2-carboxylate
Formula: C₁₀H₈INO₂S
Storage Temperature: Ambient
MDL Number: MFCD29163658
CAS Number: 2089966-09-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR