3-(3,4-Dimethoxyphenyl)propionic acid 99%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
3,4-Dimethoxyhydrocinnamic acid, 3-(3,4-Dimethoxyphenyl)propanoic acid
OR18337-100G
OR18337-25G
76569-242EA
66.22
USD
76569-242
76569-240
3-(3,4-Dimethoxyphenyl)propionic acid 99%
3-(3,4-Dimethoxyphenyl)propionic acid
Formula:
C₁₁H₁₄O₄ Melting Pt: 96…99 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002774 CAS Number: 2107-70-2 EINECS: 218-288-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...