1,4-Diethoxybenzene 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR18328-100G
OR18328-500G
OR18328-25G
76569-226EA
108.32
USD
76569-226
76569-228
76569-224
1,4-Diethoxybenzene 98%
1,4-Diethoxybenzene
Formula:
C₁₀H₁₄O₂ Boiling Pt: 245…248 °C Melting Pt: 69…72 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015146 CAS Number: 122-95-2 EINECS: 204-585-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...