4-Methoxybenzyl alcohol 98%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Anise alcohol
, 4-(Hydroxymethyl)anisole
, (4-Methoxyphenyl)methanol
, Anisyl alcohol
, p-Anisyl Alcohol
OR18325-1KG
OR18325-250G
OR18325-100G
76569-212EA
13
USD
76569-212
76569-216
76569-214
4-Methoxybenzyl alcohol 98%
4-Methoxybenzyl alcohol
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol Boiling Pt: 258…260 °C Melting Pt: 22…25 °C Density: 1.108 g/cm³ (25 °C) Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004653 CAS Number: 105-13-5 EINECS: 203-273-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...