Phenazine 99+%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
OR18320-25G
OR18320-5G
OR18320-100G
76569-204EA
26
USD
76569-204
76569-202
76569-206
Phenazine 99+%
Phenazine
Formula:
C₁₂H₈N₂ MW: 180.21 g/mol Melting Pt: 172…177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005023 CAS Number: 92-82-0 EINECS: 202-193-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...