4-Methyl-2-thio-1,3-thiazole 97%

Supplier: APOLLO SCIENTIFIC
Warning

OR18300-100G OR18300-25G
76569-150EA 103.9 USD
76569-150 76569-148
4-Methyl-2-thio-1,3-thiazole 97%
4-Methyl-2-thio-1,3-thiazole
Formula: C₄H₅NS₂
Melting Pt: 86…89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798571
CAS Number: 4498-39-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR