4-Aminoquinoline 95+%

Supplier: APOLLO SCIENTIFIC
Warning

OR17934-1G OR17934-5G
76568-944EA 28.52 USD
76568-944 76568-946
4-Aminoquinoline 95+%
4-Aminoquinoline
Formula: C₉H₈N₂
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00463448
CAS Number: 578-68-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR