2-[(Acetylthio)methyl]-3-phenylpropionic acid 96%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-[(Acetylthio)methyl]-3-phenylpropionic Acid

OR17540-5G OR17540-25G
76568-672EA 20.26 USD
76568-672 76568-674 76568-670
2-[(Acetylthio)methyl]-3-phenylpropionic acid 96%
2-[(Acetylthio)methyl]-3-phenylpropionic acid
Formula: C₁₂H₁₄O₃S
MW: 238.30 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00236751
CAS Number: 91702-98-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR