a-Tosylbenzyl isocyanide 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (1-Phenyl-1-tosyl)methyl Isocyanide, alpha-(p-Toluenesulfonyl)benzyl Isocyanide, α-Tosylbenzyl Isocyanide, α-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-(p-Toluenesulphonyl)benzyl Isocyanide, alpha-Tosylbenzyl Isocyanide, α-(p-Toluenesulfonyl)benzyl Isocyanide

OR1744-5G OR1744-1G
76568-642EA 337.18 USD
76568-642 76568-640
a-Tosylbenzyl isocyanide 97%
a-Tosylbenzyl isocyanide
Formula: C₁₅H₁₃NO₂S
MW: 271.3342 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04114776
CAS Number: 36635-66-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR