2,6-Diisopropylaniline 96%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR17130-100G
OR17130-1KG
OR17130-500G
76568-562EA
26
USD
76568-562
76568-564
76568-566
2,6-Diisopropylaniline 96%
2,6-Diisopropylaniline
Formula:
C₁₂H₁₉N MW: 177.29 g/mol Boiling Pt: 254…257 °C Melting Pt: –45 °C Density: 0.94 g/cm³ (25 °C) Flash Pt: 117 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008887 CAS Number: 24544-04-5 EINECS: 246-305-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...