3-Bromo-2-methyl-6-nitro-2H-indazole

Supplier: APOLLO SCIENTIFIC

OR16857-1G OR16857-250MG
76561-046EA 191.52 USD
76561-046 76562-612
3-Bromo-2-methyl-6-nitro-2H-indazole
3-Bromo-2-methyl-6-nitro-2H-indazole
Formula: C₈H₆BrN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD02666041
CAS Number: 74209-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR