Diphenyl phosphate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Diphenyl hydrogen phosphate

OR16436-5G OR16436-100G OR16436-25G
76568-148EA 20.32 USD
76568-148 76568-152 76568-150
Diphenyl phosphate
Diphenyl phosphate
Formula: C₁₂H₁₁O₄P
MW: 250.19 g/mol
Melting Pt: 62…70 °C
Storage Temperature: Ambient
MDL Number: MFCD00003033
CAS Number: 838-85-7
EINECS: 212-357-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR