3-(5-Bromothien-2-yl)pyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Bromo-5-(pyridin-3-yl)thiophene

OR15965-250MG OR15965-100MG OR15965-1G
76568-028EA 110.02 USD
76568-028 76568-026 76568-030
3-(5-Bromothien-2-yl)pyridine 97%
3-(5-Bromothien-2-yl)pyridine
Formula: C₉H₆BrNS
MW: 240.12 g/mol
Melting Pt: 52…54.5 °C
Storage Temperature: Ambient
MDL Number: MFCD04971290
CAS Number: 169050-05-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR