2-Pyrimidinecarboxylic acid 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pyrimidine-2-carboxylic acid, 2-Carboxypyrimidine

OR15942-25G OR15942-5G
76568-010EA 66.22 USD
76568-010 76568-008
2-Pyrimidinecarboxylic acid 97%
2-Pyrimidinecarboxylic acid
Formula: C₅H₄N₂O₂
Melting Pt: 193…200 °C
Storage Temperature: Ambient
MDL Number: MFCD00856161
CAS Number: 31519-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR