(2R,3R)-(-)-2,3-Butanediol 99%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: D(-)-2,3-Butanediol, (2R,3R)-Butane-2,3-diol, (R,R)-(-)-2,3-Butylene glykol, (R,R)-(-)-2,3-Butanediol, (2R,3R)-(-)-2,3-Dihydroxy butane, (R,R)-(-)-2,3-Dihydroxy butane, (2R,3R)-(-)-2,3-Butylene glykol

OR15918-1G OR15918-5G
76567-962EA 20.34 USD
76567-962 76567-964
(2R,3R)-(-)-2,3-Butanediol 99%
(2R,3R)-(-)-2,3-Butanediol
Formula: C₄H₁₀O₂
Boiling Pt: 179…180 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 85 °C
Storage Temperature: Ambient
MDL Number: MFCD00064267
CAS Number: 24347-58-8
EINECS: 246-186-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR