1-Bromo-2,4-dichlorobenzene

Supplier: APOLLO SCIENTIFIC
Warning

OR1562-100G OR1562-5G OR1562-25G
76567-846EA 51.96 USD
76567-846 76567-842 76567-844
1-Bromo-2,4-dichlorobenzene
1-Bromo-2,4-dichlorobenzene
Formula: C₆H₃BrCl₂
Boiling Pt: 235 °C
Melting Pt: 26…30 °C
Density: 1.89 g/cm³ (25 °C)
Flash Pt: 98 °C
Storage Temperature: Ambient
MDL Number: MFCD00018333
CAS Number: 1193-72-2
EINECS: 214-778-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR