3-Bromo-5-(hydroxymethyl)isoxazole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (3-Bromoisoxazol-5-yl)methanol

OR15590-250MG OR15590-1G
76567-834EA 48.9 USD
76567-834 76567-836
3-Bromo-5-(hydroxymethyl)isoxazole
3-Bromo-5-(hydroxymethyl)isoxazole
Formula: C₄H₄BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD04035586
CAS Number: 25742-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR