tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate 95%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR15251-1G
OR15251-500MG
OR15251-5G
76567-602EA
150.76
USD
76567-602
76567-600
76567-604
tert-Butyl-3-benzyl-2,2-dioxo-2λ~6~-diazathiane-1-carboxylate 95%
tert-Butyl-3-benzyl-2,2-dioxo-2lambda~6~-diazathiane-1-carboxylate
Formula:
C₁₂H₁₈N₂O₄S Melting Pt: 106…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD09864818 CAS Number: 147000-78-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...