3-Amino-6-methoxypyridazine

Supplier: APOLLO SCIENTIFIC
Warning

OR14965-1G OR14965-250MG
76567-318EA 65.2 USD
76567-318 76567-316
3-Amino-6-methoxypyridazine
3-Amino-6-methoxypyridazine
Formula: C₅H₇N₃O
Melting Pt: 103…108 °C
Storage Temperature: Ambient
MDL Number: MFCD01646127
CAS Number: 7252-84-8
EINECS: 230-670-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR