4-Methylquinoline
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR14907-25G
OR14907-100G
76567-282EA
110.02
USD
76567-282
76567-284
4-Methylquinoline
4-Methylquinoline
Formula:
C₁₀H₉N Boiling Pt: 261…263 °C Melting Pt: 44478 °C Density: 1.083 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006784 CAS Number: 491-35-0 EINECS: 207-734-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...