15-Hydroxypentadecanoic acid 99%

Supplier: APOLLO SCIENTIFIC
Warning

OR1489-1G OR1489-5G
76567-268EA 28 USD
76567-268 76567-270
15-Hydroxypentadecanoic acid 99%
15-Hydroxypentadecanoic acid
Formula: C₁₅H₃₀O₃
MW: 258.40 g/mol
Melting Pt: 84…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00039535
CAS Number: 4617-33-8
EINECS: 225-026-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR