L-Lysine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: (S)-2,6-Diaminohexanoic acid, L(+)-Lysine, (S)-(+)-2,6-Diaminohexanoic acid, (S)-(+)-Lysine, (S)-(+)-2,6-Diaminocaproic acid, H-Lys-OH

OR14752-1KG OR14752-100G OR14752-500G
76567-198EA 116.4 USD
76567-198 76567-194 76567-196
L-Lysine
L-Lysine
Formula: C₆H₁₄N₂O₂
MW: 146.19 g/mol
Melting Pt: 215 °C
Storage Temperature: Ambient
MDL Number: MFCD00064433
CAS Number: 56-87-1
EINECS: 200-294-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR