2-Amino-5-hydroxymethylpyridine 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Amino-5-pyridinemethanol, 6-Amino-3-pyridinemethanol, 2-Amino-5-(hydroxymethyl)pyridine, 2-Aminopyridine-5-methanol, (6-Aminopyridin-3-yl)methanol, (6-Amino-3-pyridyl)methanol

OR14503-250MG OR14503-1G
76567-088EA 22.42 USD
76567-088 76567-090
2-Amino-5-hydroxymethylpyridine 97%
2-Amino-5-hydroxymethylpyridine
Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 113…120 °C
Storage Temperature: Ambient
MDL Number: MFCD06659089
CAS Number: 113293-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR