tert-Butyl (piperazin-1-yl)acetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: tert-Butyl 2-(piperazin-1-yl)acetate, 1-[(tert-Butoxycarbonyl)methyl]piperazine, (Piperazin-1-yl)acetic acid tert-butyl ester

OR14391-250MG OR14391-1G
76567-040EA 22.42 USD
76567-040 76567-042 76567-044
tert-Butyl (piperazin-1-yl)acetate
tert-Butyl (piperazin-1-yl)acetate
Formula: C₁₀H₂₀N₂O₂
Boiling Pt: 72 °C (0.15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD06659535
CAS Number: 112257-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR