4-Cyano-4'-pentylbiphenyl 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4'-Pentyl-4-biphenylcarbonitrile, 4'-Pentyl-[1,1'-biphenyl]-4-carbonitrile
OR13991-10G
OR13991-5G
OR13991-1G
76566-640EA
131.4
USD
76566-640
76566-638
76566-636
4-Cyano-4'-pentylbiphenyl 98%
4-Cyano-4'-pentylbiphenyl
Formula:
C₁₈H₁₉N MW: 249.36 g/mol Melting Pt: 212…213/3mm °C Density: 1.008 g/cm³ (25 °C) Flash Pt: 113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00036350 CAS Number: 40817-08-1 EINECS: 255-093-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...