2-Chloro-4-methylpyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-4-picoline

OR13604-100G OR13604-500G OR13604-25G
76566-432EA 41.76 USD
76566-432 76566-434 76566-430
2-Chloro-4-methylpyridine
2-Chloro-4-methylpyridine
Formula: C₆H₆ClN
MW: 127.57 g/mol
Boiling Pt: 194…195 °C
Melting Pt: 115
Density: 1.142 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00023418
CAS Number: 3678-62-4
EINECS: 222-951-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR