4-Amino-3-nitropyridine

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Nitropyridin-4-amine, 3-Nitro-4-pyridylamine, 3-Nitropyridin-4-ylamine

OR13224-5G OR13224-25G
76566-256EA 20.26 USD
76566-256 76566-258
4-Amino-3-nitropyridine
4-Amino-3-nitropyridine
Formula: C₅H₅N₃O₂
MW: 139.11 g/mol
Melting Pt: 200…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00160304
CAS Number: 1681-37-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR