6-Cyanonicotinic acid 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR13130-250MG
OR13130-1G
76566-164EA
41.76
USD
76566-164
76566-166
6-Cyanonicotinic acid 97%
6-Cyanonicotinic acid
Formula:
C₇H₄N₂O₂ MW: 148.12 g/mol Melting Pt: 187…194 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318135 CAS Number: 70165-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...