(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 5-Amino-2-(hydroxymethyl)benzeneboronic acid dehydrate hydrochloride, 6-Aminobenzo[c][1,2]oxaborol-1(3H)-ol hydrochloride

OR13108-1G OR13108-250MG
76566-140EA 188.46 USD
76566-140 76566-138
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
(5-Amino-2-hydroxymethylphenyl)boronic acid hydrochloride
Formula: C₇H₉BClNO₂
MW: 185.42 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD04115645
CAS Number: 117098-93-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR