4-Aminophenylboronic acid pinacol ester
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-Aminobenzeneboronic acid pinacol ester
OR13050-1G
OR13050-25G
OR13050-5G
76566-078EA
20.32
USD
76566-078
76566-082
76566-080
4-Aminophenylboronic acid pinacol ester
4-Aminophenylboronic acid pinacol ester
Formula:
C₁₂H₁₈BNO₂ Melting Pt: 165…171 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02093721 CAS Number: 214360-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...