Picolinamidine hydrochloride

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Picolinimidamide hydrochloride, 2-Amidinopyridine hydrochloride

OR1303-5G OR1303-25G OR1303-1G
76566-036EA 38.72 USD
76566-036 76566-038 76566-034
Picolinamidine hydrochloride
Picolinamidine hydrochloride
Formula: C₆H₈ClN₃
Melting Pt: 145…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00052271
CAS Number: 51285-26-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR