1-Phenyl-1,3-butandione

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Phenylbutane-1,3-dione, 1-Benzoylacetone, Benzoylacetone, 1-Phenyl-1,3-butanedione

OR12814-5G OR12814-25G
76565-948EA 21.38 USD
76565-948 76565-950
1-Phenyl-1,3-butandione
1-Phenyl-1,3-butandione
Formula: C₆H₅C(O)CH₂C(O)CH₃
MW: 162.19 g/mol
Boiling Pt: 262
Melting Pt: 54…56 °C
Density: 1.09 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008786
CAS Number: 93-91-4
EINECS: 202-286-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR