trans-4-Pentylcyclohexanecarboxylic acid

Supplier: APOLLO SCIENTIFIC
Warning

OR11972-25G OR11972-5G
76565-502EA 41.76 USD
76565-502 76565-500
trans-4-Pentylcyclohexanecarboxylic acid
trans-4-Pentylcyclohexanecarboxylic acid
Formula: C₁₁H₂₀O₂
Melting Pt: 51…53 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00009964
CAS Number: 38289-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR