5-Amino-2-chlorophenol

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Chloro-5-aminophenol

OR11940-5G OR11940-25G OR11940-100G
76565-472EA 23.44 USD
76565-472 76565-474 76565-476
5-Amino-2-chlorophenol
5-Amino-2-chlorophenol
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Melting Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01707858
CAS Number: 6358-06-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR